Gene Transcripts

Gene Symbol
Bend7
Entrez Gene ID
209645
Gene Name
BEN domain containing 7
Gene Aliases
1110017O21Rik, E130319B15Rik
Chromosome Location
Chr. 2: 4717831 - 4802146 on Build GRCm38
UniGene ID
Mm.149539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190400.1 NP_001177329.1 5 1077
NM_178663.4 NP_848778.1 5 1077
GenBank mRNA AK030428.1 - 5 764
AK042822.1 - 5 1075
AK053896.1 - 5 1079
BC056476.1 - 2 125
BC096430.1 - 5 749 5041388
BC119370.1 - 5 716 8734119
BC144864.1 - 5 716 9053412

View Full Product Details