Gene Transcripts

Gene Symbol
Clrn3
Entrez Gene ID
212070
Gene Name
clarin 3
Gene Aliases
AI649392, Tmem12
Chromosome Location
Chr. 7: 135511456 - 135528654 on Build GRCm38
UniGene ID
Mm.39721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178669.5 NP_848784.1 3 648
GenBank mRNA AK033715.1 - 3 608
AK085267.1 - 3 587
AK146608.1 - 3 649
BC036176.1 - 3 636 4989703

View Full Product Details