Gene Transcripts

Gene Symbol
Iffo2
Entrez Gene ID
212632
Gene Name
intermediate filament family orphan 2
Gene Aliases
C79267
Chromosome Location
Chr. 4: 139530548 - 139620382 on Build GRCm38
UniGene ID
Mm.30464
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205173.1 NP_001192102.1 4, 5 1184
NM_183148.3 NP_898971.1 7, 8 804
GenBank mRNA BC027170.1 - 3, 4 330
BC100381.1 - 7, 8 804
BE860309.1 - 2, 3 173
CN718972.1 - 1, 2 148
EH105040.1 - 1, 2 120

View Full Product Details