Gene Transcripts

Gene Symbol
PEG3
Entrez Gene ID
5178
Gene Name
paternally expressed 3
Gene Aliases
PW1, ZKSCAN22, ZNF904, ZSCAN24
Chromosome Location
Chr. 19: 56810077 - 56840726 on Build GRCh38
UniGene ID
Hs.731875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146184.1 NP_001139656.1 8, 9 1177
NM_001146185.1 NP_001139657.1 8, 9 863
NM_001146186.1 NP_001139658.1 6, 7 1210
NM_001146187.1 NP_001139659.1 8, 9 869
NM_006210.2 NP_006201.1 9, 10 1253
GenBank mRNA AB006625.2 - 8, 9 842
AF208967.1 - 9, 10 1223
AF208968.1 - 8, 9 1147
AF208969.1 - 8, 9 833
AF208970.1 - 6, 7 1211
AK295679.1 - 8, 9 869
AK307476.1 - 8, 9 869
BC136268.1 - 7, 8 697 9020255
BC150272.1 - 8, 9 842 9007244

View Full Product Details