Gene Transcripts

Gene Symbol
Tdrd5
Entrez Gene ID
214575
Gene Name
tudor domain containing 5
Gene Aliases
Gm103
Chromosome Location
Chr. 1: 156255296 - 156305295 on Build GRCm38
UniGene ID
Mm.252800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134741.1 NP_001128213.1 7 1048
NM_001277730.1 NP_001264659.1 7 754
GenBank mRNA AK132674.1 - 7 1034
AK133026.1 - 5 419
BC099972.1 - 7 754 30080156

View Full Product Details