Gene Transcripts

Gene Symbol
Lman2l
Entrez Gene ID
214895
Gene Name
lectin, mannose-binding 2-like
Gene Aliases
A630028F14Rik, VIP36-like, VIPL
Chromosome Location
Chr. 1: 36423183 - 36445271 on Build GRCm38
UniGene ID
Mm.287854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013374.1 NP_001013392.1 5 643
GenBank mRNA AK041656.1 - 3 2488
AK142024.1 - 612
AK158015.1 - 5 662
BC027226.1 - 4 561 2654285
BC046969.1 - 5 643 6313679

View Full Product Details