Gene Transcripts

Gene Symbol
Slc36a3
Entrez Gene ID
215332
Gene Name
solute carrier family 36 (proton/amino acid symporter), member 3
Gene Aliases
PAT3, TRAMD2, tramdorin2
Chromosome Location
Chr. 11: 55124815 - 55151745 on Build GRCm38
UniGene ID
Mm.87054
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172258.3 NP_758462.2 5 601
GenBank mRNA AF453745.1 - 5 601
AK132910.1 - 6 687
AY211261.1 - 5 583

View Full Product Details