Gene Transcripts

Gene Symbol
Nav1
Entrez Gene ID
215690
Gene Name
neuron navigator 1
Gene Aliases
9530089B19, 9930003A20Rik, C230080M11Rik, Pomfil3, Unc53h1, mKIAA1151, steerin-1
Chromosome Location
Chr. 1: 135434580 - 135688116 on Build GRCm38
UniGene ID
Mm.34977
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173437.2 NP_775613.2 16, 18 3670
GenBank mRNA AF307453.1 - 16, 18 4030
CB523329.1 - 1, 3 120

View Full Product Details