Gene Transcripts

Gene Symbol
Med16
Entrez Gene ID
216154
Gene Name
mediator complex subunit 16
Gene Aliases
95kDa, A630083L04, Thrap5, Trap95
Chromosome Location
Chr. 10: 79894707 - 79908941 on Build GRCm38
UniGene ID
Mm.260089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163276.1 NP_001156748.1 15 2578
NM_198107.2 NP_932775.2 15 2581
GenBank mRNA AK042340.1 - 15 2543
AK149345.1 - 15 2562
BC057056.1 - 15 2535 6813010
DT908554.1 - 2 129

View Full Product Details