Gene Transcripts

Gene Symbol
Appl2
Entrez Gene ID
216190
Gene Name
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2
Gene Aliases
Dip3b
Chromosome Location
Chr. 10: 83600033 - 83648854 on Build GRCm38
UniGene ID
Mm.282985
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145220.2 NP_660255.1 17 1608
GenBank mRNA AK160075.1 - 17 1587
AK160105.1 - 17 1584
AK161003.1 - 14 3160
AK168403.1 - 17 1571
AK168748.1 - 17 1564
AK171512.1 - 17 1564
AK195688.1 - 3 161
AK198914.1 - 3 162
AY113706.1 - 17 1610
BC002232.1 - 5 419
BC048906.1 - 17 1546 6475576

View Full Product Details