Gene Transcripts

Gene Symbol
Ehbp1
Entrez Gene ID
216565
Gene Name
EH domain binding protein 1
Gene Aliases
AF424697, Flj21950, NACSIN
Chromosome Location
Chr. 11: 22005825 - 22287078 on Build GRCm38
UniGene ID
Mm.491265
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252515.1 NP_001239444.1 14 2372
NM_153078.4 NP_694718.4 14 2297
GenBank mRNA AF424697.1 - 13 2147
AK173055.1 - 14 2373

View Full Product Details