Gene Transcripts

Gene Symbol
Wscd1
Entrez Gene ID
216881
Gene Name
WSC domain containing 1
Gene Aliases
mKIAA0523
Chromosome Location
Chr. 11: 71749923 - 71789647 on Build GRCm38
UniGene ID
Mm.48648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177618.4 NP_808286.2 9 1775
GenBank mRNA AK220346.1 - 8 1481
BC030477.1 - 9 1912
BC048831.1 - 8 1594 4504495
BC063070.1 - 9 1775 5687975

View Full Product Details