Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145440.1 | NP_663415.1 | 1 | 352 | |
GenBank mRNA | AF441863.1 | - | 1 | 352 | |
AY065981.1 | - | 1 | 311 | ||
BC125312.1 | - | 1 | 331 | 40129827 | |
BC125318.1 | - | 1 | 331 | 40129833 |