Gene Transcripts

Gene Symbol
Ifi27l2b
Entrez Gene ID
217845
Gene Name
interferon, alpha-inducible protein 27 like 2B
Gene Aliases
1810023F06Rik, ISG12(b2)
Chromosome Location
Chr. 12: 103450898 - 103457223 on Build GRCm38
UniGene ID
Mm.298416
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145449.2 NP_663424.1 4, 5 442
GenBank mRNA AK007590.1 - 2, 3 333
BC021795.1 - 4, 5 442 4217067

View Full Product Details