Gene Transcripts

Gene Symbol
Ttc37
Entrez Gene ID
218343
Gene Name
tetratricopeptide repeat domain 37
Gene Aliases
mKIAA0372
Chromosome Location
Chr. 13: 76098730 - 76190323 on Build GRCm38
UniGene ID
Mm.277964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081352.1 NP_001074821.1 10 954
GenBank mRNA AK134255.1 - 10 900
AK164627.1 - 10 955
EF591876.1 - 566

View Full Product Details