Gene Transcripts

Gene Symbol
Rasa1
Entrez Gene ID
218397
Gene Name
RAS p21 protein activator 1
Gene Aliases
Gap, RasGAP, Rasa
Chromosome Location
Chr. 13: 85214699 - 85289493 on Build GRCm38
UniGene ID
Mm.259653
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145452.3 NP_663427.2 9 1826
GenBank mRNA AK133354.1 - 5 386
BC013637.1 - 7 572 3498774
BC065082.1 - 9 1369

View Full Product Details