Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205345.1 | NP_001192274.1 | 2 | 275 | |
NM_001205346.1 | NP_001192275.1 | 2 | 275 | ||
NM_194342.3 | NP_919323.2 | 2 | 275 | ||
GenBank mRNA | AK046318.1 | - | 2 | 244 | |
AK085196.1 | - | 2 | 244 | ||
AK128958.1 | - | 2 | 269 | ||
AK156246.1 | - | 2 | 276 | ||
AK171058.1 | - | 2 | 244 | ||
AK172398.1 | - | 2 | 276 | ||
AK172996.1 | - | 2 | 100 | ||
AY682987.1 | - | 1 | 41 | ||
BC098208.1 | - | 2 | 195 | 6827666 |