Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110017.1 | NP_001103487.1 | 26, 27 | 3279 | |
NM_027341.2 | NP_081617.1 | 25, 26 | 2661 | ||
GenBank mRNA | AK122344.1 | - | 3, 4 | 1822 | |
BC117953.1 | - | 25, 26 | 2484 | 40098141 | |
BC117954.1 | - | 25, 26 | 2490 | 40098143 | |
EU233997.1 | - | 18, 19 | 2481 |