Gene Transcripts

Gene Symbol
Acat3
Entrez Gene ID
224530
Gene Name
acetyl-Coenzyme A acetyltransferase 3
Gene Aliases
ACTL
Chromosome Location
Chr. 17: 12923959 - 12940396 on Build GRCm38
UniGene ID
Mm.490312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153151.3 NP_694791.2 6 835
GenBank mRNA AF356876.1 - 6 841
AK143884.1 - 6 795
AK149441.1 - 6 835
BC049873.1 - 6 740 5054536

View Full Product Details