Gene Transcripts

Gene Symbol
Oosp2
Entrez Gene ID
225922
Gene Name
oocyte secreted protein 2
Gene Aliases
Gm99, Plac1l, Tmem122
Chromosome Location
Chr. 19: 11647279 - 11661605 on Build GRCm38
UniGene ID
Mm.26881
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037634.3 NP_001032723.1 2, 3 291
GenBank mRNA AK143411.1 - 2, 3 292
BC099498.1 - 2, 3 251 30931566

View Full Product Details