Gene Transcripts

Gene Symbol
Dyrk3
Entrez Gene ID
226419
Gene Name
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3
Gene Aliases
BC006704
Chromosome Location
Chr. 1: 131128441 - 131138234 on Build GRCm38
UniGene ID
Mm.39299
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145508.2 NP_663483.1 3 1056
GenBank mRNA AK035114.1 - 835
AK161495.1 - 3 968
BC006704.1 - 3 1056 3584001

View Full Product Details