Gene Transcripts
- Gene Symbol
- Slc25a25
- Entrez Gene ID
- 227731
- Gene Name
- solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25
- Gene Aliases
- 1110030N17Rik, MCSC, mKIAA1896
- Chromosome Location
- Chr. 2: 32414487 - 32451470 on Build GRCm38
- UniGene ID
- Mm.37395
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164357.1 |
NP_001157829.1
|
4, 5 |
738 |
|
NM_001164358.1 |
NP_001157830.1
|
5, 6 |
993 |
|
NM_001290558.1 |
NP_001277487.1
|
4, 5 |
607 |
|
NM_146118.3 |
NP_666230.2
|
5, 6 |
774 |
|
GenBank mRNA |
AK049392.1 |
- |
4, 5 |
714 |
|
AK082756.1 |
- |
4, 5 |
695 |
|
AK122568.2 |
- |
5, 6 |
958 |
|
AK132204.1 |
- |
4, 5 |
692 |
|
BC019978.1 |
- |
4, 5 |
546 |
4239441 |
BC022114.1 |
- |
4, 5 |
544 |
5098924 |
BC037109.1 |
- |
5, 6 |
734 |
5368306 |
BC066998.1 |
- |
5, 6 |
961 |
5703622 |
View Full Product Details