Gene Transcripts

Gene Symbol
Gmeb2
Entrez Gene ID
229004
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
AI839884
Chromosome Location
Chr. 2: 181251449 - 181287974 on Build GRCm38
UniGene ID
Mm.220423
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198169.2 NP_937812.1 3 286
GenBank mRNA AB221583.1 - 2 166
AB221584.1 - 2 166
BC029763.1 - 2 109
BC059066.1 - 3 286 6825428

View Full Product Details