Gene Transcripts

Gene Symbol
Sprr4
Entrez Gene ID
229562
Gene Name
small proline-rich protein 4
Gene Aliases
Chromosome Location
Chr. 3: 92500263 - 92500493 on Build GRCm38
UniGene ID
Mm.246590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173070.1 NP_775093.1 1 178
GenBank mRNA AY158996.1 - 1 178
BC107019.1 - 1 178 40049167

View Full Product Details