Gene Transcripts

Gene Symbol
Trim45
Entrez Gene ID
229644
Gene Name
tripartite motif-containing 45
Gene Aliases
4921530N01Rik
Chromosome Location
Chr. 3: 100922493 - 100936925 on Build GRCm38
UniGene ID
Mm.189351
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165952.1 NP_001159424.1 2 939
NM_001165953.1 NP_001159425.1 2 939
NM_194343.2 NP_919324.2 2 939
GenBank mRNA AK076600.1 - 2 515
AK150574.1 - 2 940
AK153571.1 - 2 940
BC057358.1 - 2 817 6835531

View Full Product Details