Gene Transcripts

Gene Symbol
Rap1gds1
Entrez Gene ID
229877
Gene Name
RAP1, GTP-GDP dissociation stimulator 1
Gene Aliases
BC011279, GDS1
Chromosome Location
Chr. 3: 138925897 - 139075201 on Build GRCm38
UniGene ID
Mm.28893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040690.2 NP_001035780.1 9, 10 1204
NM_001286759.1 NP_001273688.1 9, 10 1204
NM_145544.3 NP_663519.2 8, 9 1057
GenBank mRNA AK052125.1 - 8, 9 1005
AK154852.1 - 9, 10 1190
AK155793.1 - 9, 10 1190
AK160989.1 - 9, 10 1140
AK164159.1 - 9, 10 1116
AK166315.1 - 9, 10 1137
AK172052.1 - 9, 10 1206
BC011279.1 - 8, 9 926 4008751
CN702379.1 - 17

View Full Product Details