Gene Transcripts
- Gene Symbol
- Foxj3
- Entrez Gene ID
- 230700
- Gene Name
- forkhead box J3
- Gene Aliases
- C330039G02Rik, Fhd6
- Chromosome Location
- Chr. 4: 119539661 - 119629119 on Build GRCm38
- UniGene ID
- Mm.259227
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290696.1 |
NP_001277625.1
|
3, 4 |
575 |
|
NM_172699.3 |
NP_766287.1
|
3, 4 |
575 |
|
GenBank mRNA |
AK082840.1 |
- |
3, 4 |
576 |
|
AK129271.1 |
- |
3, 4 |
548 |
|
AK133640.1 |
- |
3, 4 |
545 |
|
AK139508.1 |
- |
3, 4 |
509 |
|
AK163298.1 |
- |
5, 6 |
688 |
|
AK171564.1 |
- |
3, 4 |
552 |
|
BC058231.1 |
- |
3, 4 |
548 |
6314981 |
CB245622.1 |
- |
1, 2 |
256 |
|
CJ048244.1 |
- |
3, 4 |
545 |
|
View Full Product Details