Gene Transcripts

Gene Symbol
Sh2d5
Entrez Gene ID
230863
Gene Name
SH2 domain containing 5
Gene Aliases
BC036961
Chromosome Location
Chr. 4: 138250411 - 138260968 on Build GRCm38
UniGene ID
Mm.482408
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099631.1 NP_001093101.1 4 329
GenBank mRNA AK032598.1 - 4 263
BC036961.1 - 5 363
DQ832275.1 - 4 329

View Full Product Details