Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002667.4 | NP_002658.1 | 2 | 320 | |
GenBank mRNA | AK311799.1 | - | 2 | 255 | |
BC005269.1 | - | 2 | 255 | 3934199 | |
BP257094.1 | - | 2 | 320 | ||
BP259965.1 | - | 2 | 304 | ||
M60411.1 | - | 1 | 167 | ||
M63603.1 | - | 2 | 274 |