Gene Transcripts

Gene Symbol
Dok7
Entrez Gene ID
231134
Gene Name
docking protein 7
Gene Aliases
A930013K19Rik, AW049091, Dok-7, EF-12, Oit5
Chromosome Location
Chr. 5: 35056566 - 35087839 on Build GRCm38
UniGene ID
Mm.205483
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172708.3 NP_766296.1 9 1735
GenBank mRNA AK044445.1 - 9 1736

View Full Product Details