Gene Transcripts

Gene Symbol
PLRG1
Entrez Gene ID
5356
Gene Name
pleiotropic regulator 1
Gene Aliases
Cwc1, PRL1, PRP46, PRPF46, TANGO4
Chromosome Location
Chr. 4: 154534997 - 154550433 on Build GRCh38
UniGene ID
Hs.249996
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201564.1 NP_001188493.1 13 1336
NM_002669.3 NP_002660.1 13 1363
GenBank mRNA AF044333.1 - 12 1238
AK002108.1 - 13 1338
BC020786.1 - 13 1300 4809482
BC106004.1 - 13 1301 3889109

View Full Product Details