Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010825.3 | NP_001010825.2 | 3 | 1563 | |
GenBank mRNA | AK044446.1 | - | 3 | 1372 | |
AK054325.1 | - | 3 | 1328 | ||
AK087477.1 | - | 3 | 1547 | ||
BC039952.1 | - | 1 | 797 | ||
BC106090.1 | - | 3 | 1534 | ||
BC139821.1 | - | 2 | 1200 | 40141450 |