Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145210.2 | NP_660211.2 | 4 | 828 | |
GenBank mRNA | AB067531.1 | - | 4 | 802 | |
AK087823.1 | - | 4 | 828 | ||
AY055830.1 | - | 4 | 801 | ||
AY055831.1 | - | 4 | 801 | ||
BC053721.1 | - | 4 | 825 | 30053184 | |
BC094918.1 | - | 4 | 818 | 30050168 | |
BC108956.1 | - | 4 | 824 | 40046020 |