Gene Transcripts

Gene Symbol
Zfp12
Entrez Gene ID
231866
Gene Name
zinc finger protein 12
Gene Aliases
C530015C18, Krox-7, Zfp-12, Znf12
Chromosome Location
Chr. 5: 143235163 - 143248834 on Build GRCm38
UniGene ID
Mm.55211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289589.1 NP_001276518.1 5 1032
NM_001289590.1 NP_001276519.1 5 1102
NM_177681.4 NP_808349.1 4 936
GenBank mRNA AK049650.1 - 4 892
BC053080.1 - 5 941 5702966
BC059836.1 - 5 1013 5697949

View Full Product Details