Gene Transcripts

Gene Symbol
Gxylt2
Entrez Gene ID
232313
Gene Name
glucoside xylosyltransferase 2
Gene Aliases
Glt8d4, Gm152
Chromosome Location
Chr. 6: 100704293 - 100805686 on Build GRCm38
UniGene ID
Mm.272037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198612.2 NP_941014.1 4 940
GenBank mRNA BC049816.1 - 4 940

View Full Product Details