Gene Transcripts

Gene Symbol
Ankrd26
Entrez Gene ID
232339
Gene Name
ankyrin repeat domain 26
Gene Aliases
5730521P14Rik, BC008111, mKIAA1074
Chromosome Location
Chr. 6: 118501308 - 118562269 on Build GRCm38
UniGene ID
Mm.276116
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081112.1 NP_001074581.1 24 3753
GenBank mRNA AK173096.1 - 8 1940
BC008111.1 - 4 1299
BC056364.1 - 7 1812

View Full Product Details