Gene Transcripts
- Gene Symbol
- Clec2e
- Entrez Gene ID
- 232409
- Gene Name
- C-type lectin domain family 2, member e
- Gene Aliases
- Clra, clr-a
- Chromosome Location
- Chr. 6: 129091816 - 129100903 on Build GRCm38
- UniGene ID
- Mm.30870
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153506.4 |
NP_705726.3
|
2 |
142 |
|
View Full Product Details