Gene Transcripts

Gene Symbol
Prr12
Entrez Gene ID
233210
Gene Name
proline rich 12
Gene Aliases
6720469B10, BC058674
Chromosome Location
Chr. 7: 45027707 - 45052881 on Build GRCm38
UniGene ID
Mm.219137
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175022.2 NP_778187.2 4 1168
GenBank mRNA AK173130.1 - 1 607

View Full Product Details