Gene Transcripts

Gene Symbol
Xylt1
Entrez Gene ID
233781
Gene Name
xylosyltransferase 1
Gene Aliases
8030490L12
Chromosome Location
Chr. 7: 117380979 - 117667630 on Build GRCm38
UniGene ID
Mm.461427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175645.3 NP_783576.2 11 2451
GenBank mRNA AJ291750.1 - 9 1768
AJ539164.1 - 11 2453

View Full Product Details