Gene Transcripts

Gene Symbol
Dkk4
Entrez Gene ID
234130
Gene Name
dickkopf homolog 4 (Xenopus laevis)
Gene Aliases
Dkk-4
Chromosome Location
Chr. 8: 22623685 - 22627547 on Build GRCm38
UniGene ID
Mm.157322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145592.2 NP_663567.1 4 756
GenBank mRNA BC018400.1 - 4 756

View Full Product Details