Gene Transcripts

Gene Symbol
Ppan
Entrez Gene ID
235036
Gene Name
peter pan homolog (Drosophila)
Gene Aliases
A230087P06Rik, SSF1
Chromosome Location
Chr. 9: 20888175 - 20892179 on Build GRCm38
UniGene ID
Mm.2930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145610.2 NP_663585.2 3, 4 413
GenBank mRNA AK039027.1 - 3, 4 318
BC014688.1 - 3, 4 331 4489442

View Full Product Details