Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276452.1 | NP_001263381.1 | 2 | 201 | |
NM_172773.3 | NP_766361.1 | 2 | 201 | ||
GenBank mRNA | AK029102.1 | - | 2 | 184 | |
AK087395.1 | - | 2 | 184 | ||
AK169868.1 | - | 2 | 150 | ||
BC058785.1 | - | 2 | 171 | 6309785 |