Gene Transcripts

Gene Symbol
Plscr4
Entrez Gene ID
235527
Gene Name
phospholipid scramblase 4
Gene Aliases
AV245873
Chromosome Location
Chr. 9: 92457372 - 92492516 on Build GRCm38
UniGene ID
Mm.55289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178711.3 NP_848826.1 6 1021
GenBank mRNA AK035878.1 - 6 1006
AK035919.1 - 6 1022
AK045372.1 - 6 1022
AK054258.1 - 6 579
AK082330.1 - 5 464
AK141580.1 - 547
AK165919.1 - 6 1022
AK166080.1 - 6 513
BC052067.1 - 6 575 5708712

View Full Product Details