Gene Transcripts

Gene Symbol
Mettl22
Entrez Gene ID
239706
Gene Name
methyltransferase like 22
Gene Aliases
Chromosome Location
Chr. 16: 8470753 - 8490205 on Build GRCm38
UniGene ID
Mm.214953
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146247.1 NP_666359.1 5 629
GenBank mRNA AK089212.1 - 5 644
AK136614.1 - 5 655
BC024814.1 - 5 629 5359405

View Full Product Details