Gene Transcripts

Gene Symbol
Tgm6
Entrez Gene ID
241636
Gene Name
transglutaminase 6
Gene Aliases
TGM3L, TGY
Chromosome Location
Chr. 2: 130112416 - 130154232 on Build GRCm38
UniGene ID
Mm.83437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289747.1 NP_001276676.1 3 308
NM_001289748.1 NP_001276677.1 2 145
NM_001289749.1 NP_001276678.1 2 164
NM_177726.4 NP_808394.1 2 164
GenBank mRNA AK037229.1 - 2 154
AY159126.1 - 2 154
AY177606.1 - 2 154
AY177607.1 - 2 145
BB612839.1 - 2 164
BC112420.1 - 2 141 40061385
BC113796.1 - 3 285 40061392

View Full Product Details