Gene Transcripts

Gene Symbol
Bank1
Entrez Gene ID
242248
Gene Name
B cell scaffold protein with ankyrin repeats 1
Gene Aliases
A530094C12Rik, AI451642, AVIEF, BANK
Chromosome Location
Chr. 3: 136053363 - 136326141 on Build GRCm38
UniGene ID
Mm.30832
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033350.2 NP_001028522.2 4 780
GenBank mRNA AK041071.1 - 1 38
AK041242.1 - 4 741
AK156201.1 - 4 790
AY178734.1 - 4 800
BC026932.1 - 4 727 4008261
EU051378.1 - 3 381

View Full Product Details