Gene Transcripts

Gene Symbol
Cyp2j12
Entrez Gene ID
242546
Gene Name
cytochrome P450, family 2, subfamily j, polypeptide 12
Gene Aliases
BB197342, Cyp2j12-ps, OTTMUSG00000007939
Chromosome Location
Chr. 4: 96099314 - 96141272 on Build GRCm38
UniGene ID
Mm.302562
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100182.2 NP_001093652.1 1 268
GenBank mRNA AK136922.1 - 1 269

View Full Product Details