Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153591.4 | NP_705819.3 | 14 | 1658 | |
GenBank mRNA | AK046799.1 | - | 14 | 1659 | |
AK046801.1 | - | 14 | 1659 | ||
AK047091.1 | - | 14 | 1603 | ||
AK050718.1 | - | 14 | 1597 | ||
AK138260.1 | - | 14 | 1638 | ||
AK164179.1 | - | 14 | 1627 | ||
BC030041.1 | - | 10 | 862 | 1226297 |