Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198858.1 | NP_942155.1 | 3 | 381 | |
GenBank mRNA | AB125661.1 | - | 3 | 381 | |
AK145438.1 | - | 453 | |||
AY363390.1 | - | 3 | 347 | ||
BC120763.1 | - | 3 | 373 | 40129686 | |
BC120765.1 | - | 3 | 373 | 40129688 |