Gene Transcripts

Gene Symbol
Erlin2
Entrez Gene ID
244373
Gene Name
ER lipid raft associated 2
Gene Aliases
BC036333, C87251, Spfh2
Chromosome Location
Chr. 8: 27023266 - 27039437 on Build GRCm38
UniGene ID
Mm.277699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153592.2 NP_705820.1 5 383
GenBank mRNA AK030557.1 - 5 327
BC036333.1 - 5 348 3590904
BC038374.1 - 5 323 5355446

View Full Product Details